You searched for: Bottle Top Filters
Bottle-top filters are used in research laboratories for sterilization or laboratory fluid clarification. The attachments are ideal for use with cell culture and serum-containing media, and asymmetrical hydrophilic membrane styles provide high flow rates. Suitable for tissue culture applications, these filters provide low protein binding and low extractables. Reusable models offer convenience and cumulative cost savings over time, with cleaning performed via autoclaving. Plastic construction imbues these filters with reliable durability.
VWR® Vacuum Filtration Systems featuring Pall Membranes
Supplier: VWR International
VWR® Vacuum Filtration Systems are designed for the preparation of buffers, tissue culture media, microbiological media, and other biological fluids.
Expand 24 Items
VWR® Tube Top Vacuum Filters
Supplier: VWR International
Minimize unnecessary transfers by filtering directly into 50 ml centrifuge tubes.
Expand 5 Items
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
Systems for vacuum filtration of aqueous solutions including cell culture media, buffers, or other biological fluids.
Expand 72 Items
AcroCap™ Positive Pressure Devices, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroCap™ devices are presterilized, disposable filter units ideal for sterilization or clarification of up to 3 L of tissue culture media or other critical fluids.
Expand 3 Items
VacuCap® and VacuCap® PF Bottle-Top Filters, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
For sterilization or clarification of laboratory fluids. Ideal for cell culture and serum-containing media.
Expand 10 Items
Corning® Filter Top Tubes, Sterile, Corning
Supplier: Corning
Cellulose acetate membrane, PS funnel and PP collection tube.
Expand 2 Items
Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Complete filter units.
Expand 6 Items
Nalgene® Analytical Filter Units, Cellulose Nitrate, 150 ml, Thermo Scientific
Supplier: Thermo Fisher Scientific
Complete, presterilized, non-toxic, disposable filter units for membrane filter analysis.
Expand 2 Items
Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Supplier: Thermo Fisher Scientific
For easy, fast, sterile filtration of 150 to 1000 ml volumes.
Expand 12 Items
Sartoclear Dynamics® Lab V Clarification and Sterile Filtration Kits, Sartorius
Supplier: Sartorius
Sartoclear Dynamics® Lab products are filtration kits designed for the clarification and sterile filtration of up to 1000 ml mammalian cell cultures.
Expand 3 Items
Nalgene® Rapid-Flow™ Tissue Culture Filter Units, Nylon Membrane, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Units are designed for applications requiring low extractables.
Expand 10 Items
Corning® Bottle-Top Filters, Sterile, Corning
Supplier: Corning
PS bottle top vacuum filters with adapters colour coded by membrane type: Polyethersulfone (PES) yellow; cellulose acetate (CA) orange; nylon (NY) red.
Expand 15 Items
Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
Expand 3 Items
Sartolab® RF 50 Tube Top Vacuum Filtration Systems, Sartorius
Supplier: Sartorius
Sartolab® RF 50 vacuum filtration units are single-use units for the filtration of sample volumes up to 50 ml. They are available with either a 0.22 or 0.45 µm polyethersulfone membrane.
Expand 4 Items
Nalgene® Filter Storage Bottle Receivers, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
These receiver storage bottles feature an ergonomic design with tapered sides and grips for ease of handling.
Expand 4 Items
Nalgene® Analytical Test Filter Funnels, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter funnels are low-cost disposable, sterile filter funnels for microbiological analysis and QC testing, food/beverages, raw material, sterility testing and finished product quality.
Expand 2 Items
Nalgene® Analytical Filter Funnels, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Ideal for use with standard filtering flasks or manifolds.
Expand 1 Items
Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
SFCA (surfactant-free cellulose acetate) membrane filters are much cleaner than standard cellulose acetate and are designed for vacuum filtration with glass media bottles.
Expand 10 Items
Bottle, Filtering, Ace Glass Incorporated
Supplier: Ace Glass
This heavy wall, bottle shaped, filtering flask with removable polypropylene hose connection is available clear glass or plastic coated.
Expand 10 Items
Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters, PES Membrane, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
All of these filters come with an asymmetric polyethersulfone (PES) membranes.
Expand 25 Items
Instatherm® Filtration Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
This apparatus differs from the standard 47mm filter apparatus (89083-540), as it uses Ace Glass’ proprietary Instatherm® technology to evenly heat the top 400 ml funnel, thus keeping the viscous materials in a flowing liquid state.
Expand 2 Items
Nalgene® Rapid-Flow™ Tissue Culture Filter Units, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Presterilized filter units with surfactant-free cellulose acetate membranes are ideal for proteinaceous solutions.
Expand 11 Items
Sartolab® BT and RF Vacuum Filtration Units, Sartorius
Supplier: Sartorius
These bottle-top filters and vacuum filtration units are single-use units for the sterile filtration of samples.
Expand 1 Items
Nalgene® Analytical Test Filter Funnels, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
For microbiological testing of water quality, food/beverages, raw material, sterility, and finished product quality.
Expand 4 Items
Corning® Vacuum Filter/Storage Systems, Sterile, Corning
Supplier: Corning
Vacuum filter systems available in four sizes: 150 ml; 250 ml; 500 ml and 1 L. Adapters are colour coded by membrane type for easy product identification: Polyethersulfone (PES) yellow; cellulose acetate (CA) orange; cellulose nitrate (CN) blue; nylon (NY) red.
Expand 22 Items
Plastic 150 ml Tube Top Filter 50 mm Membrane, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
These tubes minimize unnecessary transfer by filtering directly into the centrifuge tube.
Expand 2 Items
Bottletop Vacuum/Filter Unit
Supplier: Corning
This convenient system features a 90 mm Ø opening to filter faster through 0.22 μm membranes where autoclaving is not required.
Expand 1 Items
Disposible Vacuum Filters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Choice of membrane media: Cellulose acetate (CA), for low protein binding.
Expand 1 Items
Autofil® 2 Bottle Top Filtration Devices, Hydrophilic PES Membrane, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
The Autofil® 2 features 0.1, 0.2, or 0.45 μm PES membranes. Perfect for filtering FBS and other viscous solutions. The membrane is low protein binding and surfactant-free.
Expand 18 Items
Vacuum Filtration Bottle Top Filter Systems, Argos Technologies
Supplier: Argos Technologies
Ideal for separation, purification, and sterilization of biological samples.