Orders LinkContactUsLinkComponent
Cryogenic 2D Matrix Vials
Cryogenic 2D Matrix Vials
Supplier:   NEST SCIENTIFIC
Cryogenic 2D Matrix Vials
Supplier:  NEST SCIENTIFIC
Certificates
Cryogenic 2D Matrix Vials
26 Options Available Below
Product Details & Documents
2D barcode cryogenic vials are designed for storage of biological specimens. Each tube is permanently marked with a unique 2D barcode, ensuring the tubes can be easily and accurately identified when being moved, shipped or even accidentally dropped.
  • Laser 2D barcode on the bottom of each tube allows scan and decode at once without removing from rack
  • 100% inspection guarantees that each code is unique and readable
  • Autoclavable
  • Clear graduations and white marking area
  • Sterilized by E-Beam
  • Non-pyrogenic, DNase/RNase and human DNA free
  • Recommend to store in gas phase of the nitrigen tank
Solid, one-piece construction ensures the barcode not fall off during the storage under low temperature.
External thread cap: Special screw cap designed without silicone ring to avoid the contamination risk.
Internal thread cap: Special screw cap designed with silicone O-ring to secure the sealing under various condition.
Refrigeration: The lowest temperature can withstand −196 °C (with screw cap). All our polypropylene pipes are suitable for low temperature storage, except for liquid nitrogen.
Caution: Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Product Family Options
External Threaded
Product Information
  • Capacity
    Bottom style
    Material
    Cap included
    Cap type
    Sterility
    Packaging
    Availability
    Price
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,042.33
    $1,042.33
    Log in for availability
    Each
    Each - $1,042.33
    $1,042.33
    Case of 2,000
       $1,042.33
    Each
       $1,042.33
    Supplier Number: 618041
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 1296/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,296
    Case of 1,296 - $813.15
    $813.15
    Log in for availability
    Each
    Each - $813.15
    $813.15
    Case of 1,296
       $813.15
    Each
       $813.15
    Supplier Number: 618053
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,185.88
    $1,185.88
    Log in for availability
    Each
    Each - $1,185.88
    $1,185.88
    Case of 2,000
       $1,185.88
    Each
       $1,185.88
    Supplier Number: 606041
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 1134/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,134
    Case of 1,134 - $744.46
    $744.46
    Log in for availability
    Each
    Each - $744.46
    $744.46
    Case of 1,134
       $744.46
    Each
       $744.46
    Supplier Number: 606053
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,640.24
    $1,640.24
    Log in for availability
    Each
    Each - $1,640.24
    $1,640.24
    Case of 2,000
       $1,640.24
    Each
       $1,640.24
    Supplier Number: 607041
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 972/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 972
    Case of 972 - $937.50
    $937.50
    Log in for availability
    Each
    Each - $937.50
    $937.50
    Case of 972
       $937.50
    Each
       $937.50
    Supplier Number: 607053
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 648/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $1,033.01
    $1,033.01
    Log in for availability
    Each
    Each - $1,033.01
    $1,033.01
    Case of 648
       $1,033.01
    Each
       $1,033.01
    Supplier Number: 624053
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,093.32
    $1,093.32
    Log in for availability
    Each
    Each - $1,093.32
    $1,093.32
    Case of 1,000
       $1,093.32
    Each
       $1,093.32
    Supplier Number: 608041
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 648/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $913.41
    $913.41
    Log in for availability
    Each
    Each - $913.41
    $913.41
    Case of 648
       $913.41
    Each
       $913.41
    Supplier Number: 608053
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $843.16
    $843.16
    Log in for availability
    Each
    Each - $843.16
    $843.16
    Case of 1,000
       $843.16
    Each
       $843.16
    Supplier Number: 609041
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 486/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 486
    Case of 486 - $614.02
    $614.02
    Log in for availability
    Each
    Each - $614.02
    $614.02
    Case of 486
       $614.02
    Each
       $614.02
    Supplier Number: 609053
Internal Threaded
Product Information
  • Capacity
    Bottom style
    Material
    Cap included
    Cap type
    Sterility
    Packaging
    Availability
    Price
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,241.90
    $1,241.90
    Log in for availability
    Each
    Each - $1,241.90
    $1,241.90
    Case of 2,000
       $1,241.90
    Each
       $1,241.90
    Supplier Number: 618141
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    100/box, 1600/case, 10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,600
    Case of 1,600 - $1,145.74
    $1,145.74
    Log in for availability
    Each
    Each - $1,145.74
    $1,145.74
    Case of 1,600
       $1,145.74
    Each
       $1,145.74
    Supplier Number: 618152
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 1296/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,296
    Case of 1,296 - $965.84
    $965.84
    Log in for availability
    Each
    Each - $965.84
    $965.84
    Case of 1,296
       $965.84
    Each
       $965.84
    Supplier Number: 618153
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,362.34
    $1,362.34
    Log in for availability
    Each
    Each - $1,362.34
    $1,362.34
    Case of 2,000
       $1,362.34
    Each
       $1,362.34
    Supplier Number: 606141
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    100/box, 1400/case, 10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,400
    Case of 1,400 - $977.70
    $977.70
    Log in for availability
    Each
    Each - $977.70
    $977.70
    Case of 1,400
       $977.70
    Each
       $977.70
    Supplier Number: 606152
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 1134/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,134
    Case of 1,134 - $922.31
    $922.31
    Log in for availability
    Each
    Each - $922.31
    $922.31
    Case of 1,134
       $922.31
    Each
       $922.31
    Supplier Number: 606153
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 2000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,895.72
    $1,895.72
    Log in for availability
    Each
    Each - $1,895.72
    $1,895.72
    Case of 2,000
       $1,895.72
    Each
       $1,895.72
    Supplier Number: 607141
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    100/box, 1200/case, 10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,200
    Case of 1,200 - $1,186.19
    $1,186.19
    Log in for availability
    Each
    Each - $1,186.19
    $1,186.19
    Case of 1,200
       $1,186.19
    Each
       $1,186.19
    Supplier Number: 607152
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 972/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 972
    Case of 972 - $1,090.25
    $1,090.25
    Log in for availability
    Each
    Each - $1,090.25
    $1,090.25
    Case of 972
       $1,090.25
    Each
       $1,090.25
    Supplier Number: 607153
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $843.38
    $843.38
    Log in for availability
    Each
    Each - $843.38
    $843.38
    Case of 1,000
       $843.38
    Each
       $843.38
    Supplier Number: 624141
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 648/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $1,092.51
    $1,092.51
    Log in for availability
    Each
    Each - $1,092.51
    $1,092.51
    Case of 648
       $1,092.51
    Each
       $1,092.51
    Supplier Number: 624153
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $772.20
    $772.20
    Log in for availability
    Each
    Each - $772.20
    $772.20
    Case of 1,000
       $772.20
    Each
       $772.20
    Supplier Number: 608141
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 648/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $1,100.09
    $1,100.09
    Log in for availability
    Each
    Each - $1,100.09
    $1,100.09
    Case of 648
       $1,100.09
    Each
       $1,100.09
    Supplier Number: 608153
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/box, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $940.85
    $940.85
    Log in for availability
    Each
    Each - $940.85
    $940.85
    Case of 1,000
       $940.85
    Each
       $940.85
    Supplier Number: 609141
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    81/box, 486/case, 9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 486
    Case of 486 - $621.67
    $621.67
    Log in for availability
    Each
    Each - $621.67
    $621.67
    Case of 486
       $621.67
    Each
       $621.67
    Supplier Number: 609153