PIG® MRO Economy Spill Kit, New Pig
Catalog # 56620-300
Supplier: New Pig
Specifications
- Description:MRO Economy Spill Kit
- Cat. no.:56620-300
- Supplier No.:KIT242
Specifications
About this item
Maintenance, repair, and operation (MRO) spill kit is packed inside a heat-sealed disposal bag and is small enough to be placed anywhere in a facility or on a transport truck
- Helps facilities comply with 40 CFR 263.30(a) safety standard
Safety-yellow bag is highly visible, labeled, and may also be reused for temporary disposal of saturated absorbents. Kit contents absorb up to 18.9L (5gal.) of nonaggressive liquids such as oils, coolants, solvents, and water.
Ordering information: Kit contains three 7.6dia.x121.9Lcm (3x48") blue socks, twelve 50.8Lx40.6Wcm (20x16") universal mat pads, and two temporary disposal bags and ties.
Customers Who Bought This Also Bought