Standard Dithizone Solution, 10 mg/L in Chloroform USP 251 for Lead
Catalog # RCR2631000500C
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:10 mg/L
- Pack type:Glass bottle
- Description:Standard dithizone solution, 10 mg/L in chloroform USP <251> for lead
- Size:500 ml
- Cat. no.:RCR2631000500C
Specifications
About this item
Standard dithizone solution 10 mg/L in chloroform.
- Conforms to preparation per USP 251 for Lead
Stock solution for analysis.
Certifications: USP 251 for Lead.