ELISA Plates
About this item
NEST ELISA plate, made of polystyrene, is a safe, reliable, and effective supporter in enzyme-linked immuno-absorbent assay (ELISA). The features of the plates are crucial for the adsorption of antigens, antibodies, and antigen-antibody complexes.
- Uniform thickness and size
- High batch-to-batch stability and low coefficient of variation (CV) value
- Both detachable and non-detachable flat-bottom plates are offered for customers to choose from based on their applications
- The letters and numbers on the frame of the plates are convenient for detection and identification in experiments
- High transparency of clear plate bottom
- Greatly improves the flexibility in diagnostic testing (Only for detachable ELISA plates)
- Each well is locked in the frame, all at the same horizontal (Only for detachable ELISA plates)
- The design of 8-well strip allows for uniform reading and washing (Only for detachable ELISA plates)
ELISA plates from NEST adopts a unique surface treatment process, allowing to obtain ideal results in ELISA tests which are extensively employed in vitro diagnostics in medical and clinical laboratories.
The plates are made of polystyrene and are designed specifically for ELISAs. They present good adsorption performance and low baseline value.
White plates are best suited for luminescence measurements as they reflect the signal instead of absorbing it and hence enhance the luminescence signals. Raw materials show good light-proof properties, which greatly reduce well-to-well signal interference.
4 Options Available Below
Product Details & Documents
NEST ELISA plate, made of polystyrene, is a safe, reliable, and effective supporter in enzyme-linked immuno-absorbent assay (ELISA). The features of the plates are crucial for the adsorption of antigens, antibodies, and antigen-antibody complexes.
- Uniform thickness and size
- High batch-to-batch stability and low coefficient of variation (CV) value
- Both detachable and non-detachable flat-bottom plates are offered for customers to choose from based on their applications
- The letters and numbers on the frame of the plates are convenient for detection and identification in experiments
- High transparency of clear plate bottom
- Greatly improves the flexibility in diagnostic testing (Only for detachable ELISA plates)
- Each well is locked in the frame, all at the same horizontal (Only for detachable ELISA plates)
- The design of 8-well strip allows for uniform reading and washing (Only for detachable ELISA plates)
ELISA plates from NEST adopts a unique surface treatment process, allowing to obtain ideal results in ELISA tests which are extensively employed in vitro diagnostics in medical and clinical laboratories.
The plates are made of polystyrene and are designed specifically for ELISAs. They present good adsorption performance and low baseline value.
White plates are best suited for luminescence measurements as they reflect the signal instead of absorbing it and hence enhance the luminescence signals. Raw materials show good light-proof properties, which greatly reduce well-to-well signal interference.
Product Family Options
Product Information
- No. of wellsMaterialColorPackagingAvailabilityPrice
- 77508-146High binding ELISA plate, detachableHigh binding ELISA plate, detachable96PSWhite frame and transparent well5 plates/pack, 50 plates/caseCase of 50$195.29Each$195.29Supplier Number: 504201
- 77508-148High binding ELISA plate, detachableHigh binding ELISA plate, detachable96PSWhite frame and black well5 plates/pack, 50 plates/caseCase of 50$133.43Each$133.43Supplier Number: 504261
- 77508-150High binding ELISA plate, detachableHigh binding ELISA plate, detachable96PSWhite frame and white well5 plates/pack, 50 plates/caseCase of 50$268.15Each$268.15Supplier Number: 504271
- 77508-206High binding ELISA plate, undetachableHigh binding ELISA plate, undetachable96PSTransparent5 plates/pack, 50 plates/caseCase of 50$156.89Each$156.89Supplier Number: 514201