Orders LinkContactUsLinkComponent
Cryogenic Vials, External or Internal Threaded
Cryogenic Vials, External or Internal Threaded
Supplier:  NEST SCIENTIFIC
Certificates
Cryogenic Vials, External or Internal Threaded
23 Options Available Below
Product Details & Documents
Cryogenic Vials are designed for convenient and secure storage. These vials are ideal for storing valuable biological samples, ensuring their integrity and viability for long-term preservation.
  • Made of USP VI polypropylene
  • Autoclave at 121 °C/15 lbs.
  • Non-pyrogenic, DNase/RNase and human DNA free
  • Sterilized by E-beam, SAL=10⁻⁶
  • Clear graduations and white marking area
  • Stored in gas-phase liquid nitrogen
External thread cap: Special screw cap designed without silicone ring to avoid risk of contamination.
Internal thread cap: Specialscrew cap designed with silicone O-ring to secure the sealing under various conditon.
Product Family Options
External threaded
Product Information
  • Capacity
    Bottom style
    Material
    Cap included
    Cap type
    Sterility
    Packaging
    Availability
    Price
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Discontinued
    Supplier Number: -
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,296
    Case of 1,296 - $747.42
    $747.42
    Log in for availability
    Each
    Each - $747.42
    $747.42
    Case of 1,296
       $747.42
    Each
       $747.42
    Supplier Number: 618003
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Discontinued
    Supplier Number: 606001
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,134
    Case of 1,134 - $693.15
    $693.15
    Log in for availability
    Each
    Each - $693.15
    $693.15
    Case of 1,134
       $693.15
    Each
       $693.15
    Supplier Number: 606003
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,052.46
    $1,052.46
    Log in for availability
    Each
    Each - $1,052.46
    $1,052.46
    Case of 2,000
       $1,052.46
    Each
       $1,052.46
    Supplier Number: 607001
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 972
    Case of 972 - $822.58
    $822.58
    Log in for availability
    Each
    Each - $822.58
    $822.58
    Case of 972
       $822.58
    Each
       $822.58
    Supplier Number: 607003
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $531.05
    $531.05
    Log in for availability
    Each
    Each - $531.05
    $531.05
    Case of 1,000
       $531.05
    Each
       $531.05
    Supplier Number: 624001
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $850.81
    $850.81
    Log in for availability
    Each
    Each - $850.81
    $850.81
    Case of 648
       $850.81
    Each
       $850.81
    Supplier Number: 624003
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Discontinued
    Supplier Number: -
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $836.40
    $836.40
    Log in for availability
    Each
    Each - $836.40
    $836.40
    Case of 648
       $836.40
    Each
       $836.40
    Supplier Number: 608003
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $727.54
    $727.54
    Log in for availability
    Each
    Each - $727.54
    $727.54
    Case of 1,000
       $727.54
    Each
       $727.54
    Supplier Number: 609001
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 486
    Case of 486 - $519.88
    $519.88
    Log in for availability
    Each
    Each - $519.88
    $519.88
    Case of 486
       $519.88
    Each
       $519.88
    Supplier Number: 609003
Internal threaded
Product Information
  • Capacity
    Bottom style
    Material
    Cap included
    Cap type
    Sterility
    Packaging
    Availability
    Price
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $877.06
    $877.06
    Log in for availability
    Each
    Each - $877.06
    $877.06
    Case of 2,000
       $877.06
    Each
       $877.06
    Supplier Number: 618101
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,600
    Case of 1,600 - $922.74
    $922.74
    Log in for availability
    Each
    Each - $922.74
    $922.74
    Case of 1,600
       $922.74
    Each
       $922.74
    Supplier Number: 618102
  • 0.5 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,296
    Case of 1,296 - $890.69
    $890.69
    Log in for availability
    Each
    Each - $890.69
    $890.69
    Case of 1,296
       $890.69
    Each
       $890.69
    Supplier Number: 618103
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Discontinued
    Supplier Number: 606101
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,400
    Case of 1,400 - $891.89
    $891.89
    Log in for availability
    Each
    Each - $891.89
    $891.89
    Case of 1,400
       $891.89
    Each
       $891.89
    Supplier Number: 606102
  • 1.2 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,134
    Case of 1,134 - $855.13
    $855.13
    Log in for availability
    Each
    Each - $855.13
    $855.13
    Case of 1,134
       $855.13
    Each
       $855.13
    Supplier Number: 606103
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,112.65
    $1,112.65
    Log in for availability
    Each
    Each - $1,112.65
    $1,112.65
    Case of 2,000
       $1,112.65
    Each
       $1,112.65
    Supplier Number: 607101
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    10×10 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,200
    Case of 1,200 - $940.03
    $940.03
    Log in for availability
    Each
    Each - $940.03
    $940.03
    Case of 1,200
       $940.03
    Each
       $940.03
    Supplier Number: 607102
  • 2.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 972
    Case of 972 - $1,009.76
    $1,009.76
    Log in for availability
    Each
    Each - $1,009.76
    $1,009.76
    Case of 972
       $1,009.76
    Each
       $1,009.76
    Supplier Number: 607103
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $577.23
    $577.23
    Log in for availability
    Each
    Each - $577.23
    $577.23
    Case of 1,000
       $577.23
    Each
       $577.23
    Supplier Number: 624101
  • 3.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $565.56
    $565.56
    Log in for availability
    Each
    Each - $565.56
    $565.56
    Case of 648
       $565.56
    Each
       $565.56
    Supplier Number: 624103
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $568.33
    $568.33
    Log in for availability
    Each
    Each - $568.33
    $568.33
    Case of 1,000
       $568.33
    Each
       $568.33
    Supplier Number: 608101
  • 4.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 648
    Case of 648 - $580.09
    $580.09
    Log in for availability
    Each
    Each - $580.09
    $580.09
    Case of 648
       $580.09
    Each
       $580.09
    Supplier Number: 608103
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    50/pack, 500/box
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $786.02
    $786.02
    Log in for availability
    Each
    Each - $786.02
    $786.02
    Case of 1,000
       $786.02
    Each
       $786.02
    Supplier Number: 609101
  • 5.0 ml
    Round bottom, self-standing
    PP
    Yes
    Screw cap
    Sterile
    9×9 racked package
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 486
    Case of 486 - $558.90
    $558.90
    Log in for availability
    Each
    Each - $558.90
    $558.90
    Case of 486
       $558.90
    Each
       $558.90
    Supplier Number: 609103
Insert caps
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • Red
    Coloured cap insert
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $102.34
    $102.34
    Log in for availability
    Each
    Each - $102.34
    $102.34
    Case of 1,000
       $102.34
    Each
       $102.34
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • Yellow
    Coloured cap insert
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $98.30
    $98.30
    Log in for availability
    Each
    Each - $98.30
    $98.30
    Case of 1,000
       $98.30
    Each
       $98.30
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • Green
    Coloured cap insert
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $98.52
    $98.52
    Log in for availability
    Each
    Each - $98.52
    $98.52
    Case of 1,000
       $98.52
    Each
       $98.52
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • Purple
    Coloured cap insert
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $98.61
    $98.61
    Log in for availability
    Each
    Each - $98.61
    $98.61
    Case of 1,000
       $98.61
    Each
       $98.61
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • White
    Coloured cap insert
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $99.02
    $99.02
    Log in for availability
    Each
    Each - $99.02
    $99.02
    Case of 1,000
       $99.02
    Each
       $99.02