Orders LinkContactUsLinkComponent
Storage Bottles, Polystyrene
Storage Bottles, Polystyrene
Supplier:   NEST SCIENTIFIC
Storage Bottles, Polystyrene
Supplier:  NEST SCIENTIFIC
Certificates
Storage Bottles, Polystyrene
4 Options Available Below
Product Details & Documents
Reciever bottles are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • High flow rates and high throughput
  • Precise graduation on the bottle wall
  • Low protein binding
  • Gamma-ray sterilization
  • Filter diameter: 250 ml volume for the 50 mm membrane diameter 500 ml volume for the 90 mm membrane diameter
  • The design of hose connector is adaptable to various hose diameters
  • Light weight and firm outer wall structure, easy to grasp
It is mainly used in tissue culture media and biological fluids for separation and purification of large volume samples. The whole set includes membrane diaphragm, a graduated, transparent polystyrene funnel connected to a receiving bottle by a connector.
The disposable vacuum filter utilizes a vacuum pump to provide pressure differentials for processing large volumes of tissue culture fluids and other laboratory fluid solutions and the volume of processed samples could be up to several liters. The filtered samples can be stored directly in sterile collection bottles.
Certifications: ISO 9001 and 13485 certified.
Product Family Options
Product Information
  • Capacity
    Height
    Ø
    Neck diameter
    Packaging
    Availability
    Price
  • 150 ml
    80 mm
    Bottom: 89.15 mm
    42 mm
    1/pack, 12/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $199.49
    $199.49
    Log in for availability
    Each
    Each - $199.49
    $199.49
    Case of 24
       $199.49
    Each
       $199.49
    Supplier Number: 346021
  • 250 ml
    105 mm
    Bottom: 88 mm
    42 mm
    1/pack, 24/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $175.22
    $175.22
    Log in for availability
    Each
    Each - $175.22
    $175.22
    Case of 24
       $175.22
    Each
       $175.22
    Supplier Number: 342021
  • 500 ml
    145 mm
    Bottom: 96 mm
    42 mm
    1/pack, 24/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $206.61
    $206.61
    Log in for availability
    Each
    Each - $206.61
    $206.61
    Case of 24
       $206.61
    Each
       $206.61
    Supplier Number: 343021
  • 1000 ml
    178.5 mm
    Bottom: 122 mm
    42 mm
    1/pack, 12/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $183.80
    $183.80
    Log in for availability
    Each
    Each - $183.80
    $183.80
    Case of 12
       $183.80
    Each
       $183.80
    Supplier Number: 344021