About this item
These syringes allow the collection, storage, transport, and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Point style 2 is a sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
An easy-to-use twist valve and a positive rear stop prevents loss of sample and plunger blowout. Fluid path includes SST, borosilicate glass, and PTFE. Not autoclavable. Gas sterilizable.
Specifications
- Catalog No:
- 60373-602
- 60373-604
- 60373-606
- 60373-608
- 60373-620
- 60373-618
- 60373-600
- 60373-610
- 60373-612
- 60373-614
- 60373-616
- Min. temperature:
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- Needle length:
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- Point style:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2