Cell Culture Chamber Slides
Supplier: NEST SCIENTIFIC
Certificates
About this item
NEST Cell culture chamber slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocyto-logical experiments.
- Convenient for microscopic observation
- Bottom materials: Borosilicate glass, thickness 1.0 to 0.1 mm, no surface treatment for glass bottom
- Chamber color: Clear/white/black
- Easy open flip for chamber and slide disassembly
- No chemical adhesives used
- Designed to prevent cross contamination, sterile
- Alphanumeric labeling
Cell culture chamber slides provide removable chambers that enable cell growth directly on the microscopic slide, convenient for staining and microscopic examination without cell transfer. Chambers with various well numbers and colors are available for different applications.
Packing trays can be used as incubation racks in CO₂ incubators.
Frequently Bought Together