Orders LinkContactUsLinkComponent
Adhesive Microscope Slide Grids
Adhesive Microscope Slide Grids
  89032-163
 :  GRID-1000
Adhesive Microscope Slide Grids
  89032-163
Adhesive Microscope Slide Grids
Adhesive Microscope Slide Grids
Adhesive Microscope Slide Grids
$131.44
Pack of 40 Case of 960
Disposable adhesive grid stickers are ideal for counting or measuring cells and estimating the size or length of microscopic or semi-micro objects.
Stickers have two side-by-side 20 x 20 mm grids spaced 1 mm apart. Use with dry or suspended specimens such as cells held in buffer under coverslips. Separate coverslips can be placed over each sticker. Compatible with different styles of microscopes. Made of polyester with acrylic adhesive, slide grids are chemically inert and stable under most laboratory conditions.
  • Description:
    Adhesive Microscope Slide Grids