Orders LinkContactUsLinkComponent
VWR® Stopcocks
VWR® Stopcocks
VWR® Stopcocks
Supplier:  VWR International
Certificates
VWR® Stopcocks
3 Options Available Below
Product Details & Documents
These stopcocks are ideal for use with liquids and gases under low pressure (operating pressure 7.0psi) or low vacuum.
  • Ideal for low-pressure use
  • HDPE body
  • Polypropylene tap
  • Straight-bored stopcocks
The HDPE body and polypropylene tap provide a smooth turning action without the use of lubricants. Stopcocks are straight bored.
Documents
Product Family Options
Product Information
  • Nominal O.D.
    Bore size
    Availability
    Price
  • 10 mm (0.39'')
    7 mm (0.28'')
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $62.75
    $62.75
    New Customer Promotional Price
    Log in for availability
    Case of 50
    Case of 50 - $313.76
    $313.76
    New Customer Promotional Price
    Pack of 10
       $62.75
    Case of 50
       $313.76
  • 13 mm (0.51'')
    10.9 mm (0.43'')
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $207.33
    $207.33
    Log in for availability
    Case of 20
    Case of 20 - $414.66
    $414.66
    Pack of 10
       $207.33
    Case of 20
       $414.66
  • 15 mm (0.59'')
    12.7 mm (0.5'')
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $347.95
    $347.95
    Log in for availability
    Case of 20
    Case of 20 - $695.90
    $695.90
    Pack of 10
       $347.95
    Case of 20
       $695.90