Process Cooling Box
About this item
This product can used in variety of cell types, including stem cells, primary cells, cell lines and yeast. The cooling box does not require any liquid or additive, all you need is placed the cool box into the ultra-low temperature refrigerator, then it can ensure that the box's has the cooling rate of −1 ℃ per minute.
- It is a very good device for all types cell preservation
- The product uses a symmetrical design with a combination on of foam and precious alloys
- No additive and no emissions characteristics
- It is an environmental friendly products
- It does not need to replace any parts, and can be permanently used
This product is easy to use, just insert the frozen tube, placed the unit in −80 ℃ freeze environment. It can greatly improve the handling efficiency and has a good survival rate of cell preservation. The sample is safer and worry free.
It holds upto 12 tubes - 1/2 ml cryogenic vials or 1.5/2 ml centrifuge tubes.
2 Options Available Below
Product Details & Documents
This product can used in variety of cell types, including stem cells, primary cells, cell lines and yeast. The cooling box does not require any liquid or additive, all you need is placed the cool box into the ultra-low temperature refrigerator, then it can ensure that the box's has the cooling rate of −1 ℃ per minute.
- It is a very good device for all types cell preservation
- The product uses a symmetrical design with a combination on of foam and precious alloys
- No additive and no emissions characteristics
- It is an environmental friendly products
- It does not need to replace any parts, and can be permanently used
This product is easy to use, just insert the frozen tube, placed the unit in −80 ℃ freeze environment. It can greatly improve the handling efficiency and has a good survival rate of cell preservation. The sample is safer and worry free.
It holds upto 12 tubes - 1/2 ml cryogenic vials or 1.5/2 ml centrifuge tubes.
Product Family Options
Product Information
- DimensionsWeightAvailabilityPrice
- 77507-404Process cooling box for 12 tubes, HexagonalProcess cooling box for 12 tubes, HexagonalSide length: 108×100 mm120 gCase of 1$528.09Each$528.09Supplier Number: 200101
- 77507-406Process cooling box for 12 tubes, squareProcess cooling box for 12 tubes, square117×117×100 mm160 gCase of 1$528.09Each$528.09Supplier Number: 200102