Serological Pipettes
Serological pipettes can be used for liquid transfer in biological experiments such as cell culture and bacterial culture.
- Made of high clarity polystyrene
- Sterilized by E-beam
- Non-pyrogenic, non-cytotoxic and non-hemolytic
- DNase/RNase free
- Polyolefin fiber filter to reduce contamination
- Edged color-coded band for identification
- Negative graduations for extra volume
Serological pipettes can be used for liquid transfer in biological experiments such as cell culture and bacterial culture.
- Made of high clarity polystyrene
- Sterilized by E-beam
- Non-pyrogenic, non-cytotoxic and non-hemolytic
- DNase/RNase free
- Polyolefin fiber filter to reduce contamination
- Edged color-coded band for identification
- Negative graduations for extra volume
Documents
Product Family Options
Individually wrapped in plastic bags
- Packed
- 1/pack, 500/box, 3000/caseCase of 3,000$909.72Each$909.72: 324003
- 1/pack, 400/box, 2400/caseCase of 2,400$784.08Each$784.08: 325003
- 1/pack, 200/box, 800/caseCase of 800$386.46Each$386.46: 326003
- 1/pack, 50/box, 500/caseCase of 800$707.84Each$707.84: 328003
- 1/pack, 200/box, 800/caseCase of 800$429.47Each$429.47: 327003
- 1/pack, 100/box, 600/caseCase of 600$995.35Each$995.35: 329003
Individually wrapped in paper-plastic bags
- Packed
- 1/pack, 500/box, 3000/caseCase of 3,000$885.48Each$885.48: 324001
- 1/pack, 400/box, 2400/caseCase of 2,400$756.52Each$756.52: 325001
- 1/pack, 200/box, 800/caseCase of 800$376.98Each$376.98: 326001
- 1/pack, 200/box, 800/caseCase of 800$418.56Each$418.56: 327001
- 1/pack, 200/box, 800/caseCase of 800$683.90Each$683.90: 328001
- 1/pack, 100/box, 600/caseCase of 600$960.40Each$960.40: 329001
- 1/pack, 50/box, 300/caseCase of 300$867.74Each$867.74: 329501
Bulk packaging
- Packed
- 25/bulk bag, 1000/pack, 3000 boxCase of 3,000$801.59Each$801.59: 324002
- 25/bulk bag, 1000/pack, 3000 boxCase of 3,000$903.67Each$903.67: 325002
- 25/bulk bag, 500/pack, 1500 boxCase of 1,500$659.47Each$659.47: 326002
- 25/bulk bag, 200/pack, 800 boxCase of 800$726.45Each$726.45: 328002
- 25/bulk bag, 100/pack, 600 boxCase of 600$1,041.50Each$1,041.50: 329002