Indicator Solution Kit, Concentrated, Hydrion®
Concentrated Indicator Solution Kit, pH 1.0 - 11.0
Catalog # 34186-839
Supplier: Micro Essential
Supplier #: UI-100
Indicator Solution Kit, Concentrated, Hydrion®
Catalog # 34186-839
Supplier: Micro Essential
CAS Number:
Jump to:
Product Details & Documents
One drop of the indicator solution in a 4ml sample will provide bright, easy-to-read colors
A polyethylene dropper bottle contains 24ml of indicator solution. Includes capped 4ml sample tube and color chart packaged in a compact polypropylene case.
A color chart is furnished with each kit covering the 1–11 pH range in steps of 1 pH unit. It permits estimates to the nearest 0.5 pH unit.
Specifications
Specifications
- Description:Concentrated Indicator Solution Kit
- Range:pH 1.0 - 11.0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



