Nalgene® Syringe Filters, Cellulose Acetate, 4 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
For cold sterilization of aqueous solutions, biological or immunological samples, and cell culture media components.
- Membrane diameter: 4 mm
- Sample volume size: 0.5 to 1.0 ml
- Polypropylene housing
- Inlet: female Luer-lock
- Outlet: male Luer slip
- Non sterile
For cold sterilization of aqueous solutions, biological or immunological samples, and cell culture media components.
Excellent for small-volume assays such as EIA and ELISA. Cellulose acetate membrane is low protein binding with low extractables.
Customers Who Bought This Also Bought