ContactUsLinkComponent
Whatman™ Nylon Membrane Circles
Whatman™ Nylon Membrane Circles
Supplier:  Cytiva
Certificates
undefined
Whatman™ Nylon Membrane Circles
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • CA28159-746
  • CA28159-751
  • CA28159-743
  • CA28159-744
  • CA28159-723
  • CA28159-728
  • CA76540-416
  • CA28159-721
  • CA28159-720
  • CA28159-768
  • Max. operating temperature:
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • Application:
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions
  • Filtration of aqueous and organic mobile phases
    Vacuum degassing
    Filtration of tissue culture media, microbiological media, buffers, and solutions