Order Entry
Canada
ContactUsLinkComponent
 
Whatman™ Anodisc Circle with Support Ring, Whatman products (Cytiva)
 :  Cytiva
 
undefined
Whatman™ Anodisc Circle with Support Ring, Whatman products (Cytiva)
 :  Cytiva
 

 

  • Catalog No:
  • CA28138-069
  • CA28138-071
  • CA28138-063
  • CA28138-067
  • CA28138-073
  • CA28138-065
  • Max. operating temperature:
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • Application:
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods formation
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods fo
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods fo
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods formation
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods formation
  • HPLC mobile phase filtration and degassing
    Ultra cleaning of solvents
    Gravimetric analysis
    Liposome extrusion
    Scanning electron microscopy studies
    Bacterial analysis by epifluorescence light microscopy
    Micrometer and nanometer filtration
    Metal nanorods formation
  • Burst strength:
  • 4.48 bar min. to 7.58 bar max.
  • 4.48 bar min. to 7.58 bar max.
  • 4.48 bar min. to 7.58 bar max.
  • 4.48 bar min. to 7.58 bar max.
  • 4.48 bar min. to 7.58 bar max.
  • 4.48 bar min. to 7.58 bar max.
  • Porosity:
  • Min. 25 - max. 50%
  • Min. 25 - max. 50%
  • Min. 25 - max. 50%
  • Min. 25 - max. 50%
  • Min. 25 - max. 50%
  • Min. 25 - max. 50%
  • Protein:
  • Low adsorption
  • Low adsorption
  • Low adsorption
  • Low adsorption
  • Low adsorption
  • Low adsorption
  • Refractive index:
  • 1.6
  • 1.6
  • 1.6
  • 1.6
  • 1.6
  • 1.6

 

Frequently Bought Together