Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Columns
Accucore™ VDX C18 HPLC Column
Accucore™ VDX C18 HPLC Column
Accucore™ VDX C18 HPLC Column
Catalog # 76287-900
Supplier:
Thermo Scientific
Expand image
Accucore™ VDX C18 HPLC Column
Catalog # 76287-900
Supplier:
Thermo Scientific
Supplier Number:
VDX-102130
Specifications
Ø int.:
2.1 mm
Particle Size:
2.6 µm
Length:
100 mm
Environmentally Preferable:
Cat. no.:
76287-900
Phase:
C18
Specifications
About this item
Product Details
Product Details