You searched for: Mills
Designed for rapid, high-speed grinding of small samples, these mills utilize stainless steel or hard-faced blades to successfully pulverize even the most difficult materials. These blades either pulse or provide continuous operation depending on the specific samples. The mills had safety features that protect the operators during procedures. With powerful mechanical force, the mills work to break down solids to be uniformly sized or simply able to be studied more closely.
SP Bel-Art MICRO-MILL® II Grinding Mill, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
The MICRO-MILL® II Grinder is ideal for rapid, high-speed milling of small samples from 20 to 50 mL.
Expand 1 Items
Vial Crushers, Thermo Scientific
Supplier: Thermo Scientific
Unit breaks and separates liquid from vials and ampules for environmentally responsible waste handling.
Expand 1 Items
Alsto JC5030 Jaw Crusher
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
The Alsto JC5030 is the largest jaw crusher. A high capacity unit suitable for laboratory use and plant applications. Rugged construction and high reduction capabilities together with low maintenance requirements make it outstanding for a large variety of applications.
Expand 2 Items
Alsto Laboratory PV Mill Pulveriser
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Modern design, complete with incorporated advanced electronics ensures smooth, problem free operation. Touch screen electronics and PLC programmed pulverising cycle maximizes efficiency of operation. Pneumatic lid clamping system for a range of bowls, and an automatic door locking system preventing access to moving elements during the grinding cycle, maximizes safety.
Expand 4 Items
Alsto JC2013 Jaw Crusher
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
The Alsto JC2013 is a reliable, heavy duty crusher designed for general laboratory applications, ready and able to crush a large variety of sample types quickly and efficiently.
Expand 2 Items
Alsto JC3020 Jaw Crusher
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
The Alsto JC3020 is suitable for either laboratory use or plant applications. Rugged construction and high reduction capabilities together with low maintenance requirements make it outstanding for a large variety of applications.
Expand 2 Items
Alsto DP2000 Disc Pulveriser
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Simple, robust machine designed for fine grinding of a large variety of sample and ore types. One fixed and one rotating 200 mm grinding plate turning on a horizontal axis, with the sample drawer located at the bottom of the chamber.
Expand 2 Items
Tube Mill 100 Control, IKA
Supplier: IKA Works
The Tube Mill 100 control is a batch mill with disposable grinding chambers. Eliminates possibility of cross-contamination and the effort of cleaning.
Expand 1 Items
Polymix® Laboratory Grinding Mills, PX-MFC 90 D, Kinematica
Supplier: Kinematica
Grinding mill with powerful motor, which is suitable for all laboratory dry milling applications.
Expand 2 Items
Tube Mill Control, IKA
Supplier: IKA Works
The Tube Mill control is a highly versatile small batch mill with disposable grinding chambers there by eliminating cross-contamination and cleaning effort.
Expand 1 Items
GRINDOMIX Knife Mill, GM 300, Retsch
Supplier: Retsch
The GRINDOMIX GM 300 Knife mill is the ideal instrument for the grinding and homogenization of foodstuffs. It allows to process sample volumes up to 4.5 l quickly and reproducibly.
Expand 1 Items
Planetary Ball Mills, Retsch®
Supplier: Retsch
These ball mills pulverize and mix soft, medium-hard, and extremely hard as well as brittle and fibrous materials
Expand 2 Items
Jar Mill, General Utility, U.S. Stoneware
Supplier: US Stoneware
This compact, “roller-type” jar mill has a small footprint
Expand 1 Items
Drive Rolls-Batch Mills, Sepor
Supplier: SEPOR CA
This rugged unit is widely used in metallurgical laboratories for grinding, cyanidation, amalgamation, mixing, and other conditioning.
Expand 3 Items
Ball Mills, PTA, Nidec Shimpo
Supplier: Nidec Shimpo America
Compact design that is ideal for small-scale production and is constructed using powder-coated steel for long-lasting durability.
Expand 2 Items
Cole-Parmer® BM-600 Planetary Mill High-Energy Ball Mill
Supplier: SPEX
The Cole-Parmer® BM-600 planetary mill high-energy ball mill grinds samples with a mechanical motion rotating the jar in one direction while the platform, planetary wheel, often referred to as the sun wheel, is rotated in the opposite direction. The mechanical milling is performed at a 2:1 ratio causing the jar to rotate twice for every single rotation of the sun wheel. As the jar is moved, the relative centrifugal force is transferred to the grinding balls causing them to move in a circular motion, into each other and against the jar wall, grinding the sample. Functionally described as a high-shearing or a high-energy ball mill, this mill is used for pulverizing rocks, minerals, sand, cement, slag, ceramics, glass, and other brittle to hard samples to analytical fineness, blending powders or making emulsions.
Expand 17 Items
Big Boyd Primary Crusher with Front Loader, Rocklabs
Supplier: ROCKLABS (W/T) CA
The Boyd Crusher is the world's first double acting, fine jaw crusher
Expand 1 Items
Universal Grinding Mills, IKA® Works
Supplier: IKA Works
Suitable for grinding up to 250mL of hard, brittle substances
Expand 2 Items
Cole-Parmer® BM-400/ BM-450 Mixer/Mill® High-Energy Ball Mills
Supplier: SPEX
Easily pulverize hard or brittle samples to analytical fineness in minutes.
Expand 20 Items
Cole-Parmer® SM-200 ShatterBox® Versatile Ring and Puck Mill
Supplier: SPEX
Versatile Ring and Puck Mill with soundproof enclosure keeps noise level down while grinding samples.
Expand 11 Items
Batch Ball and Rod Mills, Sepor
Supplier: SEPOR CA
Designed to be companion equipment to the SEPOR drive rolls, these durable containers will grind from 1000 to 2500 g charges.
Expand 4 Items
CRM-RSD Continuous Flow Ring Mill Combo with Built-In Rotating Sample Divider, Rocklabs
Supplier: ROCKLABS (W/T) CA
The Continuous Flow Ring Mill is suitable for pulverizing large samples from 1kg to 10kg
Expand 2 Items
Analytical Grinding Mill, A11, IKA® Works
Supplier: IKA Works
Universal grinding mill with air-tight grinding chamber and interchangeable blades grinds a variety of substances, wet or dry
Expand 2 Items
Microfine Grinding Mill, MF 10, IKA® Works
Supplier: IKA WORKS, INC. MX
This continuous microfine, stainless steel grinder features interchangeable grinding heads
Expand 1 Items
Bench Top Rotating Sample Divider (BTRSD), Rocklabs
Supplier: ROCKLABS (W/T) CA
The Bench Top Rotating Sample Divider is designed to be placed on a work bench, It can also be used in conjunction with a Bench Top Ring Mill.
Expand 1 Items
Vibratory Pulverizer, Bico Braun International
Supplier: Bico
For efficient grinding of production samples
Expand 1 Items
Model LM2, Laboratory Ring Mills, FLSMIDTH INC.
Supplier: FLSMIDTH INC CA
Reliable vibratory mill with the power to rapidly pulverize ores, minerals, metallurgical samples, ceramics, soils, aggregates, chemicals, and similar particulates for analysis.
Expand 1 Items
Cole-Parmer® Freezer/Mill® Cryogenic Grinders
Supplier: SPEX
Grind the toughest samples at cryogenic temperatures.
Expand 8 Items
Terminator Jaw Crusher, T.M. Engineering
Supplier: TM ENGINEERING CA
The new Terminator jaw crusher features an opening adjustment on one side, using only two screws
Expand 1 Items
Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mills
Supplier: SPEX
Grind your samples with minimal loss in a soundproof enclosure.