You searched for: Beakers
Graduated and non-graduated beakers allow contents to be efficiently mixed, analyzed, and transported safely. The standard or low-form cylindrical vessels have a reinforced rim typically with a spout for mess-free pouring. With heat and chemical resistance, the containers offer maximum control even under strenuous conditions and procedures. Available in a variety of capacities and manufacturing materials, the laboratory beakers may possess additional attributes including color-coding, non-slip handles, and no-drip spouts.
VWR® Heavy-Duty Low Form Beakers
Supplier: VWR International
These beakers are mechanically stronger than standard beakers and are designed for longer life with increased user safety.
Expand 6 Items
VWR® Beakers without Handle, PP
Supplier: VWR International
VWR® polypropylene beakers, which come in a natural color, have a pouring spout and graduations printed directly on the beakers.
Expand 9 Items
VWR® TPX™ Beakers with Handle
Supplier: VWR International
VWR® TPX™ Beakers, which are crystal-clear in color, have a handle, pouring spout and graduations printed directly on the beakers.
Expand 6 Items
Kartell® Graduated Beakers, Low Form, PMP
Supplier: VWR International
Crystal-clear beakers feature excellent chemical resistance and a large spout for efficient pouring.
Expand 8 Items
VWR® Thermotech® Beakers, PTFE
Supplier: VWR International
These unique beakers are a combination of a pure PTFE body with a stabilized PTFE carbon base to form a single molding.
Expand 3 Items
Standard-Grade Glass Beaker
Supplier: VWR International
Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.
Expand 12 Items
VWR®, Graduated Beakers, Polypropylene
Supplier: VWR International
Beakers feature a pouring spout for quick and easy dispensing.
Expand 8 Items
VWR® Tall Form Beaker
Supplier: VWR International
High Quality at a Great Value
Expand 6 Items
VWR® Standard Standard-Grade Beakers
Supplier: VWR International
A Great Value!
Expand 13 Items
VWR®, Griffin Beakers, Polypropylene
Supplier: VWR International
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
Expand 9 Items
VWR® Beakers with Handle, PP
Supplier: VWR International
VWR® polypropylene beakers, which come in a natural color, have a handle, pouring spout and graduations printed directly on the beakers.
Expand 7 Items
VWR®, Beaker Cups, Polystyrene
Supplier: VWR International
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
Expand 6 Items
VWR® Berzelius Tall Form Beakers
Supplier: VWR International
Our new high-quality borosilicate Berzelius beakers meet ASTM standard and are a great value.
Expand 7 Items
VWR®, Beakers, PTFE
Supplier: VWR International
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
Expand 13 Items
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Supplier: VWR International
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
Expand 6 Items
Graduated Polypropylene Pitchers
Supplier: VWR International
These pitchers are made with shatter-proof polypropylene.
Expand 6 Items
VWR® Heavy-Duty-Grade Beakers
Supplier: VWR International
0.5 mm Thicker Wall Than Standard Grade Beakers
Expand 5 Items
VWR® Low Form Griffin Beakers
Supplier: VWR International
These low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass.
Expand 15 Items
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Supplier: VWR International
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
Expand 5 Items
VWR® Low Form Heavy Duty Beakers
Supplier: VWR International
Our high-quality borosilicate beakers meet ASTM standards at a great value.
Expand 6 Items
VWR® TPX™ Beakers without Handle
Supplier: VWR International
VWR® TPX™ Beakers, which are crystal-clear in color, have a pouring spout and graduations printed directly on the beakers.
Expand 8 Items
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Supplier: Therapak, LLC
This wide-mouth cup is ideal for initial collection of urine samples and subsequent pour off into transport vials.
Expand 1 Items
Beakers, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
This jacketed beaker has one upper and one lower hose connection on opposite sides
Expand 9 Items
SP Wilmad-LabGlass Jacketed Beakers, SP Industries
Supplier: SP Industries
Made of borosilicate glass, these laboratory beakers are fully jacketed to allow temperature control over a wide range.
Expand 2 Items
Beakers, Pilot Plant, Ace Glass Incorporated
Supplier: Ace Glass
This pilot plant beaker is used for large sizes for batch operations and mixing large volumes of measure liquids
Expand 4 Items
Beaker Mug, Graduated, Ace Glass Incorporated
Supplier: Ace Glass
This is a 400mL heavy-wall, graduated beaker mug
Expand 1 Items
Beakers, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
This jacketed beaker has one upper and one lower Ace-Thred connection on opposite sides.
Expand 9 Items
KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
These berzelius, tall form, double capacity scale beakers are manufactured with uniform wall thickness, particularly in the critical radius point of the heel area.
Expand 11 Items
Nalgene® PFA Beakers with Handles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene® PFA Beakers with Handles feature extraordinary chemical resistance and can be used repeatedly for high-purity applications
Expand 2 Items
Beakers, Quartz, Ace Glass Incorporated
Supplier: Ace Glass
This low form, quartz beaker has a pouring spout