Orders LinkContactUsLinkComponent
Pipette Tips
Supplier:   NEST SCIENTIFIC
Pipette Tips
Supplier:  NEST SCIENTIFIC
Certificates
7 Options Available Below
Product Details & Documents
NEST 5 and 10 ml pipette tips are precision-molded from high purity, DNase RNase and pyrogen free, virgin polypropylene.
  • Autoclavable at 121 ℃
  • Meet a wider range of volumetric pipetting needs for more precise pipetting
They are available in 5 and 10 ml, filtered or non-filtered. Choose from compatible pipettors either for Eppendorf, Sartorius, Gilson, or for Thermo pipettors.
They allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
 : ISO Class 08.
Product Family Options
5 ml tips
Product Information
  • Tip style
    Volume
    Sterile
    Filter
    Tip color
    For use with
    Packed
    Availability
    Price
  • Graduated
    5 ml
    -
    Clear
    For Thermo
    100/pack, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - 281,29 $
    281,29 $
    Log in for availability
    Each
    Each - 281,29 $
    281,29 $
    Case of 1,000
       281,29 $
    Each
       281,29 $
    Supplier Number: 307021
  • Graduated
    5 ml
    +
    +
    Clear
    For Eppendorf, Sartorius, Watson, Gilson
    240/pack, 1000/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 720
    Case of 720 - 620,13 $
    620,13 $
    Log in for availability
    Each
    Each - 620,13 $
    620,13 $
    Case of 720
       620,13 $
    Each
       620,13 $
    Supplier Number: 307013
  • Graduated
    5 ml
    +
    Clear
    For Thermo
    400/pack, 720/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,200
    Case of 1,200 - 647,58 $
    647,58 $
    Log in for availability
    Each
    Each - 647,58 $
    647,58 $
    Case of 1,200
       647,58 $
    Each
       647,58 $
    Supplier Number: 307022
  • Graduated
    5 ml
    +
    +
    Clear
    For Thermo
    400/pack, 1200/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,200
    Case of 1,200 - 894,35 $
    894,35 $
    Log in for availability
    Each
    Each - 894,35 $
    894,35 $
    Case of 1,200
       894,35 $
    Each
       894,35 $
    Supplier Number: 307023
10 ml tips
Product Information
  • Tip style
    Volume
    Sterile
    Filter
    Tip color
    For use with
    Packed
    Availability
    Price
  • Graduated
    10 ml
    +
    +
    Clear
    For Eppendof, Sartorius, Gilson
    240/pack, 720/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 720
    Case of 720 - 764,14 $
    764,14 $
    Log in for availability
    Each
    Each - 764,14 $
    764,14 $
    Case of 720
       764,14 $
    Each
       764,14 $
    Supplier Number: 307113
  • Graduated
    10 ml
    +
    Clear
    For Thermo
    240/pack, 720/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 720
    Case of 720 - 565,35 $
    565,35 $
    Log in for availability
    Each
    Each - 565,35 $
    565,35 $
    Case of 720
       565,35 $
    Each
       565,35 $
    Supplier Number: 307122
  • Graduated
    10 ml
    +
    +
    Clear
    For Thermo
    240/pack, 720/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 720
    Case of 720 - 757,25 $
    757,25 $
    Log in for availability
    Each
    Each - 757,25 $
    757,25 $
    Case of 720
       757,25 $
    Each
       757,25 $
    Supplier Number: 307123