ABS Plastic Indicator for Industial Applications, Defender 6000
Catalogue # 77435-060
Fournisseur: Ohaus
Spécifications
- Model:INDICATOR DT61PW
- Description:ABS plastic indicator for industial applications, Defender 6000
- Cat. no.:77435-060
Spécifications
À propos de cet article
Washdown bench ABS plastic indicator for standard industrial applications. Defender 6000 indicator constructed of polycarbonate stands up to harsh cleaning in processing and packaging environments with IP68 immersion/IP69K washdown protection.
- Large, bright backlit LCD clearly displays results for fast processing
- The color bar can be used for quick checking of package weights for fast 'go/no-go' alert
- Durably constructed with a 304 stainless steel platform, frame, IP67 loadcell, Defender 6000 hybrid meets NTEP/Measurement Canada approvals
The Defender 6000 series Hybrid Washdown bench scale is ideal for a multitude of applications, including production and packaging. Defender 6000 handles the most demanding applications with a polycarbonate housing equipped with a 3-color check light bar, ultra-long battery life and EasyConnectTM. The Defender 6000 hybrid is designed to simplify demanding industrial and commercial applications.