Order Entry
Canada
ContactUsLinkComponent
 
BioBasic™ PREP SEC 60 HPLC Columns
  76420-886
BioBasic™ PREP SEC 60 HPLC Columns
  76420-886
 :  73305-307846A

 

  • Column Format:
    Preparative
  • Particle Size:
    5 µm
  • Pore size:
    60 Å
  • Max. pressure:
    4300 psi
  • Length:
    300 mm
  • Endcapped:
    No
  • USP code:
    L59
  • pH range:
    2 - 8
  • Surface area:
    450 m²/g
  • Temperature limit:
    60 °C
  • Carbon load:
    >9%
  • Cat. no.:
    76420-886
  • Packing Material:
    Spherical, fully porous, ultrapure silica
  • Phase:
    Diol (OH)

 

 

BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 60,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases

Stationary phase: Diol (OH)