Carboy Caddy, Labconco®
360 lbs., 20", 23⅝", 38½", 21 lbs.
Catalog # 82006-858
Supplier: Labconco
Supplier #: 8000300
Jump to:
Product Details & Documents
Low-profile cart simplifies storage and delivery of carboys, jugs, and large reagent bottles
Cumbersome glassware and equipment slide easily on to the 10.6cm (43/16") high platform. Glacier white epoxy-coated 18-gauge tubular steel construction supports up to 163kg (360lbs.) on 7.6cm (3") hard rubber casters. Features a melamine laminate platform surface that is chemically and thermally resistant and a handle with an ergonomic extruded rubber grip.
Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled.
Documents
Labconco FlaskScrubber and SteamScrubber Laboratory Glassware Washers
Product Literature
Specifications
- Width:23⅝"
- Height:38½"
- Length:20"
- Load capacity:360 lbs.
- Weight:21 lbs.



