Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Silanized Sample Vial

Silanized vial, 12 ml, 19×65 mm, Borosilicate glass, Amber, Yes, 15-425, 200 Pk
  66065-612
 :  B7999-S12A
Silanized Sample Vial
  66065-612
Silanized Sample Vial

Silanized vial, 12 ml, 19×65 mm, Borosilicate glass, Amber, Yes, 15-425, 200 Pk

 
$451.48
 
   
Pack of 200
 

Improve sample analysis sensitivity by eliminating sample/glass reactions.

  • Superior quality borosilicate clear or amber glass
  • Eliminate leaching of ions
  • Provide consistent pH for duration of sample storage life
 
  • Cap included:
    Yes
  • Cap Size:
    15-425
  • Capacity:
    12 ml
  • Color:
    Amber
  • Description:
    Silanized vial
  • Material:
    Borosilicate glass
  • Packaging:
    200 Pk
  • Diameter ×H:
    19×65 mm