Orders LinkContactUsLinkComponent
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histological solvents. Cassettes keep specimens safely submerged in liquid while the efficient flow-through slots maximize fluid exchange and ensure proper drainage.
Ordering information: Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Product Family Options
  • Color
  • White
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-2
  • Pink
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-3
  • Green
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-4
  • Yellow
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-5
  • Blue
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-6
  • Peach
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-7
  • Tan
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-8
  • Gray
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-9
  • Lilac
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-10
  • Orange
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-11
  • Aqua
    Case of 1,500
    Case of 1,500 - $496.53
    $496.53
    Case of 1,500
       $496.53
     : M515-12
  • Fluorescent Pink
    Case of 1,500
    Case of 1,500 - $496.54
    $496.54
    Case of 1,500
       $496.54
     : M515-16
  • Fluorescent Green
    Case of 1,500
    Case of 1,500 - $496.54
    $496.54
    Case of 1,500
       $496.54
     : M515-17
  • Fluorescent Yellow
    Case of 1,500
    Case of 1,500 - $496.54
    $496.54
    Case of 1,500
       $496.54
     : M515-18
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)