Orders LinkContactUsLinkComponent
UNISETTE™ Processing and Embedding Cassettes, Simport
UNISETTE™ Processing and Embedding Cassettes, Simport
Supplier:  Simport Scientific
Certificates
UNISETTE™ Processing and Embedding Cassettes, Simport
UNISETTE™ Processing and Embedding Cassettes, Simport
24 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The disposable biopsy cassettes have 1mm square openings and are specially designed to hold biopsy specimens during the embedding process, as well as in a storage file.
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through slots allow for maximum fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one-handed operation. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle.
 : Cassettes are available in 11 noncytotoxic, nonmetallic colors, and three fluorescent colors. Each case contains three dispenser boxes of 500 cassettes.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • CA73960-548
    Standard Cassettes
    Standard Cassettes
    Fluorescent Pink
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-548
  • CA73960-550
    Standard Cassettes
    Standard Cassettes
    Fluorescent Green
    Log in for availability
    Log in for availability
    Case of 1,500
    Case of 1,500 - $395.70
    $395.70
    Case of 1,500
       $395.70
    Supplier Number: M505-17
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-550
  • CA73960-552
    Standard Cassettes
    Standard Cassettes
    Fluorescent Yellow
    Log in for availability
    Log in for availability
    Case of 1,500
    Case of 1,500 - $395.70
    $395.70
    Case of 1,500
       $395.70
    Supplier Number: M505-18
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-552
  • CA73960-554
    Biopsy Cassettes
    Biopsy Cassettes
    Fluorescent Pink
    Log in for availability
    Log in for availability
    Case of 1,500
    Case of 1,500 - $404.61
    $404.61
    Case of 1,500
       $404.61
    Supplier Number: M506-16
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-554
  • CA73960-556
    Biopsy Cassettes
    Biopsy Cassettes
    Fluorescent Green
    Log in for availability
    Log in for availability
    Case of 1,500
    Case of 1,500 - $404.61
    $404.61
    Case of 1,500
       $404.61
    Supplier Number: M506-17
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-556
  • CA73960-558
    Biopsy Cassettes
    Biopsy Cassettes
    Fluorescent Yellow
    Log in for availability
    Log in for availability
    Case of 1,500
    Case of 1,500 - $404.61
    $404.61
    Case of 1,500
       $404.61
    Supplier Number: M506-18
    Specifications:
    More
    Specifications
    Cat. No.
    CA73960-558
  • CA60830-098
    Standard Cassettes
    Standard Cassettes
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.96
    $141.96
    Log in for availability
    Case of 1500
    Case of 1500 - $425.88
    $425.88
    Pack of 500
       $141.96
    Case of 1500
       $425.88
    Supplier Number: M505-2
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-098
  • CA60830-100
    Standard Cassettes
    Standard Cassettes
    Pink
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-3
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-100
  • CA60830-102
    Standard Cassettes
    Standard Cassettes
    Green
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.30
    $141.30
    Log in for availability
    Case of 1500
    Case of 1500 - $423.90
    $423.90
    Pack of 500
       $141.30
    Case of 1500
       $423.90
    Supplier Number: M505-4
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-102
  • CA60830-104
    Standard Cassettes
    Standard Cassettes
    Yellow
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.96
    $141.96
    Log in for availability
    Case of 1500
    Case of 1500 - $425.88
    $425.88
    Pack of 500
       $141.96
    Case of 1500
       $425.88
    Supplier Number: M505-5
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-104
  • CA60830-106
    Standard Cassettes
    Standard Cassettes
    Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.96
    $141.96
    Log in for availability
    Case of 1500
    Case of 1500 - $425.88
    $425.88
    Pack of 500
       $141.96
    Case of 1500
       $425.88
    Supplier Number: M505-6
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-106
  • CA60830-108
    Standard Cassettes
    Standard Cassettes
    Peach
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-7
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-108
  • CA60830-110
    Standard Cassettes
    Standard Cassettes
    Tan
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-8
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-110
  • CA60830-112
    Standard Cassettes
    Standard Cassettes
    Gray
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-9
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-112
  • CA60830-114
    Standard Cassettes
    Standard Cassettes
    Lilac
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-10
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-114
  • CA60830-116
    Standard Cassettes
    Standard Cassettes
    Orange
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-116
  • CA60830-118
    Standard Cassettes
    Standard Cassettes
    Aqua
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.92
    $142.92
    Log in for availability
    Case of 1500
    Case of 1500 - $428.76
    $428.76
    Pack of 500
       $142.92
    Case of 1500
       $428.76
    Supplier Number: M505-12
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-118
  • CA60830-120
    Biopsy Cassettes
    Biopsy Cassettes
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $158.61
    $158.61
    Log in for availability
    Case of 1500
    Case of 1500 - $475.83
    $475.83
    Pack of 500
       $158.61
    Case of 1500
       $475.83
    Supplier Number: M506-2
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-120
  • CA60830-122
    Biopsy Cassettes
    Biopsy Cassettes
    Pink
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.63
    $141.63
    Log in for availability
    Case of 1500
    Case of 1500 - $424.89
    $424.89
    Pack of 500
       $141.63
    Case of 1500
       $424.89
    Supplier Number: M506-3
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-122
  • Biopsy Cassettes
    Green
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-124
  • CA60830-126
    Biopsy Cassettes
    Biopsy Cassettes
    Yellow
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.59
    $142.59
    Log in for availability
    Case of 1500
    Case of 1500 - $427.77
    $427.77
    Pack of 500
       $142.59
    Case of 1500
       $427.77
    Supplier Number: M506-5
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-126
  • CA60830-128
    Biopsy Cassettes
    Biopsy Cassettes
    Blue
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-128
  • CA60830-130
    Biopsy Cassettes
    Biopsy Cassettes
    Peach
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $160.46
    $160.46
    Log in for availability
    Case of 1500
    Case of 1500 - $481.38
    $481.38
    Pack of 500
       $160.46
    Case of 1500
       $481.38
    Supplier Number: M506-7
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-130
  • CA60830-132
    Biopsy Cassettes
    Biopsy Cassettes
    Tan
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $141.63
    $141.63
    Log in for availability
    Case of 1500
    Case of 1500 - $424.89
    $424.89
    Pack of 500
       $141.63
    Case of 1500
       $424.89
    Supplier Number: M506-8
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-132
  • CA60830-134
    Biopsy Cassettes
    Biopsy Cassettes
    Gray
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.59
    $142.59
    Log in for availability
    Case of 1500
    Case of 1500 - $427.77
    $427.77
    Pack of 500
       $142.59
    Case of 1500
       $427.77
    Supplier Number: M506-9
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-134
  • CA60830-136
    Biopsy Cassettes
    Biopsy Cassettes
    Lilac
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.59
    $142.59
    Log in for availability
    Case of 1500
    Case of 1500 - $427.77
    $427.77
    Pack of 500
       $142.59
    Case of 1500
       $427.77
    Supplier Number: M506-10
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-136
  • CA60830-138
    Biopsy Cassettes
    Biopsy Cassettes
    Orange
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $142.59
    $142.59
    Log in for availability
    Case of 1500
    Case of 1500 - $427.77
    $427.77
    Pack of 500
       $142.59
    Case of 1500
       $427.77
    Supplier Number: M506-11
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-138
  • CA60830-140
    Biopsy Cassettes
    Biopsy Cassettes
    Aqua
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $159.39
    $159.39
    Log in for availability
    Case of 1500
    Case of 1500 - $478.17
    $478.17
    Pack of 500
       $159.39
    Case of 1500
       $478.17
    Supplier Number: M506-12
    Specifications:
    More
    Specifications
    Cat. No.
    CA60830-140
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.