You searched for: Conductivity Sensors
Conductivity sensors are becoming increasingly important for the determination of impurities in water or the concentration measurement of dissolved chemicals. Different models meet a variety of specific functions. Many of these sensors are intended for low-conductivity applications typically found in the semi-conductor, power, water, and pharmaceutical industries, providing accurate measurements even under extreme conditions in a conveniently compact instrument. Designs facilitate shielding during immersion measurements in various aqueous and non-aqueous media.
Orion™ Three-Electrode Cells, Thermo Scientific
Supplier: Thermo Fisher Scientific
Measure samples with the utmost versatility using this 3-electrode conductivity cell, which has high, standard and low conductivity measuring ranges.
Expand 1 Items
CDC566T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
The Radiometer CDC566T Conductivity Cell is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), robust epoxy body and integrated temperature sensor
Expand 1 Items
IntelliCAL™ CDC401 Four-Poles Graphite Conductivity Cell, Hach
Supplier: Hach
The IntelliCAL™ CDC401 is a digital, graphite, 4-pole conductivity cell with a temperature sensor
Expand 1 Items
CDC511T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
The Radiometer CDC511T is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and TPX body
Expand 1 Items
CDC741T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
The Radiometer CDC741T is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), glass body and integrated temperature sensor
Expand 1 Items
Orion™ DuraProbe™ Four-Electrode Conductivity Cells, Thermo Scientific
Supplier: Thermo Fisher Scientific
Virtually eliminate fringe field interference errors with the advanced Orion™ DuraProbe™ 4-Electrode conductivity cells.
Expand 3 Items
CDC861T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
The Radiometer CDC861T is a 4-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and glass body
Expand 1 Items
CDC641T 2-pole Platinum Conductivity Cell by Radiometer Analytical, Hach
Supplier: Hach
The Radiometer Analytical CDC641T conductivity cell is a 2-pole conductivity cell with platinum sensor
Expand 1 Items
Orion™ Two-Electrode Conductivity Cells, Thermo Scientific
Supplier: Thermo Fisher Scientific
Perform applications in either the laboratory or the field with versatile Orion™ Two-Electrode Conductivity Cells, available in a variety of materials and measurement ranges.
Expand 9 Items
InLab® Conductivity Probes, Mettler Toledo
Supplier: Mettler Toledo
Flow cells are recommended for measuring conductivity in pure water or in samples with low conductivity to rule out the falsification of results by carbon dioxide.
Expand 8 Items
Conductivity Electrodes for HQd Meter, Hach
Supplier: Hach
IntelliCAL CDC401 is a digital, graphite, 4-pole conductivity probe
Expand 2 Items
Conductivity Electrode, LE703, Mettler-Toledo
Supplier: Mettler Toledo
Robust conductivity probe for medium to high conductivities with integrated temperature sensor (10 μS/cm–200 mS/cm).
Expand 1 Items
LE703 Four-Pole Graphite Conductivity Probe, METTLER TOLEDO®
Supplier: Mettler Toledo
General-purpose probe is designed for use with METTLER TOLEDO® conductivity meters in a laboratory setting.
Expand 1 Items
Conductivity Cells, WTW
Supplier: WTW
This selection of conductivity cells includes the TetraCon® series of cells, which feature the highest degree of precision and linearity by optimized cell geometry, an extremely large measuring range with just one cell, long-term cell constant stability with high-quality abrasion-resistant graphite electrodes, and a built-in temperature sensor
Expand 8 Items
Conductivity Cells, IDS, for Multi 3410 / 3420 / 3430, WTW
Supplier: WTW
High quality, enhanced sensor technology combined with most modern measurement electronics.
Expand 3 Items
Conductivity Cells, YSI 3400 Series
Supplier: YSI
For use with YSI conductive instruments as well as with instruments of other manufacturers.
Expand 5 Items
Jenway Conductivity Cells for Conductivity Meters, Antylia Scientific
Supplier: Argos Technologies
These Conductivity Cells can be used with Jenway Portable Conductivity Meter (model 470) and/or Jenway Bench Conductivity Meter (model 4510 and 4520).
Expand 4 Items
MultiLab IDS Conductivity and Temperature Sensors, Two Electrodes, YSI
Supplier: YSI
The IDS sensors automatically store their unique serial number and calibration data
Expand 1 Items
MultiLab IDS Conductivity and Temperature Sensors, Four-Granite Electrodes, YSI
Supplier: YSI
The IDS sensors automatically store their unique serial number and calibration data.
Expand 2 Items
Conductivity Cells, YSI
Supplier: YSI
For use with YSI 3200 series conductivity meters.