ContactUsLinkComponent
Nalgene® 489 Low-Density Polyethylene Tubing, Thermo Scientific
undefined
Nalgene® 489 Low-Density Polyethylene Tubing, Thermo Scientific
  • Catalog No:
  • 63006-089
  • Model:
  • 63006-089
  • Max. pressure:
  • 118 psig @ 23 °C (73 °F), 59 psig @ 63 °C (145 °F)
Frequently Bought Together