BD BBL™ Streptocard™ Strep Grouping Kits, BD Diagnostics
Catalog #: CA90003-182
Supplier: BD
CAS Number:
Specifications
- Description:Streptocard Enzyme Latex Kit
- Cat. no.:CA90003-182
- Supplier No.:240950
- No. of Tests:50 Tests
Specifications
About this item
Use for the qualitative identification of Lancefield streptococcal groups
Kits will work for stretopcoccal acid groups A, B, C, F, and G and enzyme groups A, B, C, D, F, and G. Each kit provides maximum antigen yield using the enzyme extraction procedure for highly sensitive results. Each kit contains enough materials for 50 tests.
The blue latex reaction allows for easy interpretation. The reagents are packaged in color-coded dropper bottles for easy organization in the included workstation.
Customers Who Bought This Also Bought