ContactUsLinkComponent
Riffle-Type Sampler, Humboldt
Riffle-Type Sampler, Humboldt
Catalog # 56723-206
Small Business Enterprise
undefined
Riffle-Type Sampler, Humboldt
Catalog # 56723-206
Supplier Number:  H-3975
Small Business Enterprise
Specifications
  • Description:
    Riffle-Type Sampler
  • Cat. no.:
    56723-206
  • Supplier No.:
    H-3975
Specifications
About this item
For facilitating rapid and even splitting of samples of cements, ores, coke, and other dry substances.
Material poured into the top is divided by twenty-four 1Wx11.4L cm (3/8x41/2") rectangular openings and halved into representative samples until an accurate sample is secured
The smooth, stainless steel inner surface prevents residues from clinging. Stainless steel exterior. Dimensions: 38.1Hx29.2Wx35.6D cm (15x111/2x14"). Shipping weight: 22.7kg (50lbs).
Made of 18- and 20-gauge sheet steel. To retain dust, door panels are held tightly by a continuous piano hinge at the top and cabinet lock at the bottom. The two drawers are welded one-piece construction.
Certifications: Meets ASTM D271.