En Core® Sampler Cartridges, C&G Containers
89094-206
Small Business Enterprise
- Description:En Core® Sampler Cartridges
- Cat. no.:89094-206
- Supplier No.:QSSC89080010
Single-use volumetric sampling cartridges may be used with T-handle device (not included) to collect, store, and deliver 5g (0.2oz.) aliquot samples of soil type matrices
Cartridges are made of inert composite polymer and have an air-tight sealing cap to prevent loss of volatiles and to eliminate the need for chemical preservatives while in the field. Cartridges are supplied with envelope with field label. Each case contains Certificate of Analysis with lot number for maximum traceability.