Orders LinkContactUsLinkComponent
Drum Thieves, Wheaton®, DWK Life Sciences
Drum Thieves, Wheaton®, DWK Life Sciences
Drum Thieves, Wheaton®, DWK Life Sciences
Certificates
Drum Thieves, Wheaton®, DWK Life Sciences
Drum Thieves, Wheaton®, DWK Life Sciences
2 Options Available Below
Product Details & Documents
Conventional open-end drum thieves
Insert empty tube in drum, hold your thumb over top and withdraw. Soda-lime glass units easily broken for quick disposal. All are 1.09 m long.
Product Family Options
Product Information
  • Capacity
    Ø
    Availability
    Price
  • Glass
    25 ml
    0.63 cm
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $372.30
    $372.30
    New Customer Promotional Price
    Case of 100
       $372.30
    Supplier Number: 885300
  • Glass
    75 ml
    1.3 cm
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $179.35
    $179.35
    New Customer Promotional Price
    Case of 24
       $179.35
    Supplier Number: 885302