Orders LinkContactUsLinkComponent
LPG Sample Cylinders, Koehler
LPG Sample Cylinders, Koehler
Supplier:   Koehler
LPG Sample Cylinders, Koehler
Supplier:  Koehler
Certificates
LPG Sample Cylinders, Koehler
LPG Sample Cylinders, Koehler
5 Options Available Below
Product Details & Documents
Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)
Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).
 : Conform to specifications: ASTM D1265, GPA 2140.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 150 ml
    Log in for availability
    Log in for availability
    Each
    Each - $5,087.23
    $5,087.23
    Each
       $5,087.23
    Supplier Number: K27851
  • 300 ml
    Log in for availability
    Log in for availability
    Each
    Each - $5,210.83
    $5,210.83
    Each
       $5,210.83
    Supplier Number: K27852
  • 500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $5,508.99
    $5,508.99
    Each
       $5,508.99
    Supplier Number: K27853
  • 1000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $10,453.12
    $10,453.12
    Each
       $10,453.12
    Supplier Number: K27854
  • 3000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $10,486.23
    $10,486.23
    Each
       $10,486.23
    Supplier Number: K27856-00000