About this item
Kits provide a wide array of devices to collect and transport urine samples.
Transfer straws can draw urine into an evacuated tube from specimen cups or pediatric collection bags to eliminate the need for pouring. The urine collection cup has an integrated transfer device that eliminates exposure to sample and the need for pouring. The cup is graduated to indicate 120 ml (4.5 oz.) of urine. Individual 16×100 mm UA tubes have either a round bottom and a 10 ml draw or a conical bottom and an 8 ml draw. The Luer-Lock™ access device provides the security of a threaded, locking luer connection in place of a luer slip device. It is also compatible with a female luer connection of needleless IV site designed for luer lock access.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number89007-088_9087966 89007-088_9240596 89007-088_9256450 89007-088_9275708 95041-138_9220417
Conformity declaration
VT367943