You searched for: Reagent Reservoirs
Single and multi-well reagent reservoirs safely contain the required liquids in designs to accommodate all pipettor types. These containers feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner to make operations simple. Models that are compatible with robotic arms feature convoluted designs to minimize dead volume and have baffles to reduce splashing. The resistant reagent reservoirs can work efficiently in both automated and manual procedures.
VWR® 12-Channel Reagent Reservoirs
Supplier: VWR International
These 12-channel reservoirs are made from high-quality, chemical resistant PP.
Expand 2 Items
VWR® Disposable Pipetting Reservoirs
Supplier: VWR International
VWR® Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width.
Expand 17 Items
Reagent Reservoirs, Gilson
Supplier: GILSON, INC.
These reagent reservoirs are ideal for dispensing reagents and solutions using a multichannel pipette. The disposable, white PS reservoirs are available in 25-mL and 50-mL sizes.
Expand 2 Items
Axygen® Disposable Reagent Reservoirs, Corning
Supplier: Corning
These disposable reagent reservoirs are sterile and certified nonpyrogenic
Expand 12 Items
Reagent Reservoirs, 12 Channel, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Non sterile reagent reservoir made from PS.
Expand 1 Items
Costar® Reagent Reservoirs, Polystyrene, Sterile, Corning
Supplier: Corning
White PS, sterile, disposable.
Expand 4 Items
WHEATON® All Purpose Reagent Reservoirs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
These reservoirs are particularly well suited for single and multichannel pipetting. Various shape, material and capacity provides ideal solutions for many applications.
Expand 3 Items
Corning® Costar® Reagent Reservoirs, for Transtar-96® Liquid Handling System, Corning
Supplier: Corning
White PS, sterile, disposable.
Expand 3 Items
Pipetting Reservoirs, Argos Technologies
Supplier: Argos Technologies
These reagent reservoirs are created to work with 8- and 12-channel pipettes
Expand 8 Items
Matrix™ Reagent Reservoirs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Matrix™ Reagent Reservoirs are sterile and offer an extra-wide base to add rigidity and stability, helping to avoid spills.
Expand 1 Items
Axygen® Single- and Multi-Well Reservoirs, Corning
Supplier: Corning
These polypropylene reagent vessels are appropriate for automated and manual applications.
Expand 24 Items
Pipettor Reagent Reservoirs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Reagent reservoirs are designed for maximum recovery of liquid reagents.
Expand 4 Items
Reagent Reservoirs, Thermo Scientific
Supplier: Thermo Scientific
Reagent reservoirs provide a variety of solutions to suit all pipetting procedures.
Expand 3 Items
Disposable Automation Reservoirs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Disposable Automation Reservoirs are of polypropylene construction and are designed for simple, economical handling of reagents on automated systems
Expand 5 Items
Conical Reservoirs, Zymo Research
Supplier: Zymo Research
Reservoirs are available in 15 or 50 mL sizes.
Expand 2 Items
Reagent Reservoirs, 12-Channel, Argos Technologies
Supplier: Argos Technologies
These pipetting basins are designed for serial dilutions or pipetting different reagents when utilizing pipettors with up to a 12-channel capacity.
Expand 2 Items
Nalgene® Disposable Polypropylene Robotic Reservoirs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene Disposable Polypropylene Robotic Reservoirs come with either a convoluted bottom design or a flat bottom design.
Expand 4 Items
Preparative Funnel, Chromatography, Ace Glass Incorporated
Supplier: Ace Glass
Bulb type reservoir preparative funnel for chromatography preparative work.
Expand 4 Items
Micro Gas Collecting Reservoir, Ace Glass Incorporated
Supplier: Ace Glass
Gas collecting reservoir, described in Microscale Organic Laboratory, Mayo, Pike, Butcher text, as a means of collecting and sampling gaseous products when using a capillary gas delivery tube
Expand 1 Items
Reservoir, Graduated Glass, Reagent, Ace Glass Incorporated
Supplier: Ace Glass
This heavy-wall borosilicate glass bottle has three PTFE valves with Tefzel keys.
Expand 6 Items
SP Wilmad-LabGlass Flash Chromatography Reservoirs, SP Industries
Supplier: SP Industries
These reservoirs are used when larger volumes are required to elute samples from flash chromatography column.
Expand 1 Items
KIMBLE® ULTRA-WARE® Flat Bottom HPLC Reservoirs with Graduations, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Designed for preparation, storage and delivery of all liquid chromatography mobile phases. As part of our ULTRA-WARE® series, these reservoirs are recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents.
Expand 7 Items
ULTRA-WARE® Conical Bottom HPLC Reservoir, Plastic-Coated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
ULTRA-WARE® Reservoirs have been specially designed for the preparation, storage and delivery of all liquid chromatography mobile phases.
Expand 13 Items
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This versatile reservoir is ideal for use when filling multi-tube blocks or microwell plates.
Expand 1 Items
Biomek® Modular Reservoirs, Beckman Coulter®
Supplier: Beckman Coulter
The modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling system.
Expand 9 Items
Buffer Reservoir, Cytiva
Supplier: Cytiva
Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 Medium, LabMate buffer reservoir makes it easier and more convenient to equilibrate PD-10 columns.
Expand 1 Items
Reagent Reservoirs, Disposable, Sorenson Bioscience
Supplier: Sorenson Bioscience
HTS Reagent Reservoirs are made up of FDA grade polystyrene which is used for use in high throughput screening.
Expand 2 Items
Eppendorf Reagent Reservoir
Supplier: Eppendorf
Convenient autoclavable reservoir.
Expand 1 Items
BRAND Reagant Reservoir, BrandTech
Supplier: Brandtech
This reagent reservoir is made from heavy-duty polypropylene to withstand autoclaving.
Expand 3 Items
Reagent Reservoir, 5 ml, Sterile
Supplier: Heathrow Scientific
Reagent Reservoir, 5 ml, Sterile