Pierce Miser™ Western Blotting Detection Reagents, Antibody Extender Solution NC, Thermo Scientific
Catalog # CAPI32110
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Antibody Extender Solution NC
- Size:500 mL
- Environmentally Preferable:
- Cat. no.:CAPI32110
- For Membrane Area:20 Mini Blots or 1600 cm² Nitrocellulose Membrane
- Supplier No.:32110
Specifications
About this item
Pierce Miser™ Antibody Extender Solution NC is a post-transfer nitrocellulose membrane-treatment reagent that enables Western blot detection with much less primary antibody.
- Use at least three times less primary antibody than normal and retain comparable or better signal detectability
- Easy to perform, 10-minute protocol
- Works with both colorimetric and chemiluminescent detection modes and with HRP and AP systems
The solution will reduce the antibody used by 3 – 100X, while maintaining the signal strength. It is compatible with colorimetric, chemiluminescent, HRP, and AP systems.