TexNotes® Self-Adhesive Note Pads, Texwipe®
7.6 x 10.2 cm (3 x 4")
TWTX5820
: Texwipe
: TX5820
Ideal for leaving notes on documents, reports, instruments, and most cleanroom surfaces
The low contamination, polymeric elastomer adhesive allows TexNotes to be attached and removed without high residue. Cleanroom packaged. 72 sheets/pad. Color: pastel blue
- W×L:7.6 x 10.2 cm (3 x 4")