Order Entry
Canada
Orders LinkContactUsLinkComponent
Aqua-Mount® Mounting Medium, Thermo Scientific

Aqua-Mount® Mounting Medium, Thermo Scientific

Aqua-Mount® Mounting Medium, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
Aqua-Mount® Mounting Medium, Thermo Scientific
Aqua-Mount® Mounting Medium, Thermo Scientific

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

All Lerner reagents are manufactured using strictly-controlled processing procedures to ensure batch-to-batch consistency

  • Non-Fluorescing, Aqueous Formula for Immunostaining

ISO-9000 certified quality procedures include verification of product quality through actual tissue testing. Dyes used are all certified by the Biological Stain Commission.

Documents

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 41799-008
    Aqua-Mount® Mounting Medium
    Aqua-Mount® Mounting Medium
    20 mL
    Discontinued
    Supplier Number: 13800
  • CA41799-008
    Aqua-Mount® Mounting Medium
    Aqua-Mount® Mounting Medium
    20 mL
    Log in for availability
    Log in for availability
    Each (20ml)
    Each (20ml) - $101.59
    $101.59
    Each (20ml)
       $101.59

    Supplier Number: 13800

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.