Terminal Deoxynucleotidyl Transferase (TdT), EMD Millipore
Catalog # CA80500-112
Supplier: MilliporeSigma
CAS Number:
Specifications
- Size:0.155 ml
- Enzyme name:Terminal Deoxynucleotidyl Transferase (TdT)
- Cat. no.:CA80500-112
- Supplier No.:PF060-.155ML
Specifications
About this item
Biological Activity: Contains 775 units of TdT; one unit is the amount of enzyme required to transfer 1.0nmol of dAMP from dATP to the 3' -OH terminus of d(A)50 in 60 minutes at 37 °C.