Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:3000 U
- Protein/peptide name:Caspase-8
- Purity:>=90% by SDS-PAGE
- Formulation:In 500mM NaCl, 150mM imidazole, 20mM Tris-HCl, 5mM DTT, 1mM EDTA, 10% glycerol, 0.1% CHAPS, pH 7.9.
- Cat. no.:CA80050-862
- Supplier No.:218769-3000U
Specifications
About this item
In 500mM NaCl, 150mM imidazole, 20mM Tris-HCl, 5 mM DTT, 1mM EDTA, 10% glycerol, 0.1% CHAPS, pH 7.9
Member of the ICE family of cysteine proteases that has been implicated as the apical signaling protease in Fas-induced apoptosis. Suitable for the study of enzyme regulation and kinetics, target substrate cleavage, and inhibitor screening. Provided as a highly active recombinant enzyme from human cDNA expressed in E. coli. Specific activity: >=5.0x106units/mg protein.