ContactUsLinkComponent
SP Bel-Art Vacuum Chamber and Plate, Bel-Art Products, a part of SP
  13197-270
undefined
SP Bel-Art Vacuum Chamber and Plate, Bel-Art Products, a part of SP
  13197-270
 :  F42043-0000
  • Volume:
    0.2 cu. ft.
  • Size:
    22.8dia. x 16H cm (9 x 6¼")
  • Cat. no.:
    13197-270
  • Supplier No.:
    420430000
This vacuum chamber is useful for vacuum drying, degassing, and teaching demonstrations.
  • Polypropylene stopcock construction
  • TFE resin plug
  • Clear polycarbonate vacuum chamber construction
  • ABS plastic plate
The chamber top is molded of clear polycarbonate for full visibility and will withstand a full vacuum at room temperature. A strong ABS plastic plate with an O-ring seals against the chamber’s flange. The polypropylene stopcock with Teflon® TFE resin plug accepts 6.4 mm (1/4") I.D. tubing.