ContactUsLinkComponent
Blot Paper, NuSep®
Blot Paper, NuSep®
Catalog # 16003-896
Supplier:  NuSep
Blot Paper, NuSep®
Catalog # 16003-896
Supplier:  NuSep
Supplier Number:  RB-300
Specifications
  • Type:
    Sheet
  • Description:
    For Wet Blots
  • Dimensions:
    7 x 8.5 cm (23/4 x 311/32")
  • Cat. no.:
    16003-896
  • Supplier No.:
    RB-300
Specifications
About this item
Thin paper for wet blots and thick paper for semi-dry blots is 7x8.5cm (23/4x35/16").