ContactUsLinkComponent
Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Catalog # CAPI21050
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Catalog # CAPI21050
Supplier:  Thermo Scientific
Supplier Number:  21050
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Western Blot Signal Enhancer
  • Size:
    500 mL
  • Cat. no.:
    CAPI21050
  • Supplier No.:
    21050
Specifications
About this item
Western blot signal enhancer is used to pre-treat either nitrocellulose or PVDF membranes for a 3 – 10X increase in signal intensity for most target proteins.
  • Protein targets can be detected at lower levels
  • Enhances antibody binding to immobilized antigens
  • Works with any substrate, enhancing both chemiluminescence and colorimetric detection
  • Works with any membrane
  • Ready-to-use and can be stored at room temperature
Signal enhancement has been demonstrated with targets such as IL-6, p53, NF[kappa]B, BRCA1, and EGF.