ContactUsLinkComponent
SP Bel-Art Secador® 4.0 Horizontal Profile Desiccator Cabinets, Bel-Art Products, a part of SP
SP Bel-Art Secador® 4.0 Horizontal Profile Desiccator Cabinets, Bel-Art Products, a part of SP
Certificates
undefined
SP Bel-Art Secador® 4.0 Horizontal Profile Desiccator Cabinets, Bel-Art Products, a part of SP
Specifications
  • Catalog No:
  • 47743-460
  • Exterior dimensions:
  • 51.8W×41.4D×34.1H cm (20.4×16.3×13.4")
  • Int. volume:
  • 53.8 L (1.9 cu.ft.)