ContactUsLinkComponent
PIG® SpillBlocker® Tabletop Dike, New Pig
PIG® SpillBlocker® Tabletop Dike, New Pig
Catalog # 46000-742
Supplier:  New Pig
undefined
PIG® SpillBlocker® Tabletop Dike, New Pig
Catalog # 46000-742
Supplier:  New Pig
Supplier Number:  PLR229
Specifications
  • Dimensions:
    2.5 x 121.9 x 1.9 cm (1" x 4' x 0.75")
  • Cat. no.:
    46000-742
  • Supplier No.:
    PLR229
Specifications
About this item
Contains benchtop spills without absorbing; Excellent for small spills on smooth work surfaces.
  • Polyurethane material resists water, oils and many chemicals
  • High-visibility yellow color draws attention to spill for increased safety
  • Reusable dike cleans easily with soap and water
  • Comes in compact storage box for easy transport
  • Cut to any length for a custom fit
Can also be used to form an edge on workbenches to keep bolts, bearings and other small parts from rolling off.