SP Bel-Art Spill Containment Tray with Grid, Bel-Art Products, a part of SP
Catalog # 74950-156
Supplier: Bel-Art Products, a Part of SP
Specifications
- Volume:7.0 L (237 oz.)
- Cat. no.:74950-156
- Exterior Dimensions:36.5L x 30.8W x 12.1H cm (143/8 x 121/8 x 43/4")
- Supplier No.:246760003
Specifications
About this item
Large volume benchtop containment device protects users by capturing accidental or hazardous spills.
- Epoxy-coated steel wire support grid
- Molded polypropylene tray
- Excellent chemical resistance
- Autoclavable
An epoxy-coated wire support grid suitable for holding numerous vessels or tube racks is set within the large, deep polypropylene molded tray. Tray and grid have excellent chemical resistance. Steam autoclavable at 121°C (250°F).