ContactUsLinkComponent
PIG® Spill Containment Tray, New Pig
PIG® Spill Containment Tray, New Pig
  46000-750
 :  New Pig
undefined
PIG® Spill Containment Tray, New Pig
  46000-750
 :  New Pig
 :  PAK371
  • Dimensions:
    30.5 x 42.2 x 9.9 cm (12 x 16.63 x 3.88")
  • Environmentally Preferable:
  • Cat. no.:
    46000-750
  • Sump Capacity:
    5.7 L (1.5 gal.)
  • Supplier No.:
    PAK371
Chemical-resistant polyethylene tray features built-in sump containment to trap leaks and drips while you work.
  • Catch spills when containers leak, spill, or crack
  • Ideal for storing small containers on lab countertop
  • Chemical-resistant polyethylene trays have built-in containment reservoir
  • Translucent sidewalls make leaks visible
  • Pour spout allows easy draining
Translucent sidewalls let you easily inspect fluid level in sump. Compact size fits on tabletop or counter; Excellent for use in labs. Pour spout for easy draining.