ContactUsLinkComponent
Synthware Hickman Distillation Head, with Side Port, Microscale, Kemtech America
Catalog # 60004-138
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Hickman Distillation Head, with Side Port, Microscale, Kemtech America
Catalog # 60004-138
Supplier:  Kemtech America
Supplier Number:  H121410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. no.:
    60004-138
  • ST Joint Size:
    14/10
  • Supplier No.:
    H121410
Specifications
About this item
Hickman still head with side port for syringe access.
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
  • Hickman Syle still head with side port
For microscale fractional distillation. Supplied with a nylon compression cap, Viton* O-ring, and side port cap with 8 mm PTFE-lined silicone septum.