ContactUsLinkComponent
Synthware Condenser, Coil Reflux, Microscale, Kemtech America
Synthware Condenser, Coil Reflux, Microscale, Kemtech America
Catalog # 60004-126
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Condenser, Coil Reflux, Microscale, Kemtech America
Catalog # 60004-126
Supplier:  Kemtech America
Supplier Number:  C321410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • ST Joint:
    14/10
  • Cat. no.:
    60004-126
  • Supplier No.:
    C321410
Specifications
About this item
Microscale coil reflux condenser for low boiling solvents.
  • Heavy wall Borosilicate construction
  • Threaded Upper joint
  • Open cap included
Supplied with a nylon compression cap and Viton® O-ring.